Product Details

SNP ID
rs17112801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:106489247 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGACCCAGTTCAGTGGTTCCTAA[G/T]TACACCCTGGTGACCTGATCCTTCC
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IGH PubMed Links

Gene Details

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

Gene
LINC00221
Gene Name
long intergenic non-protein coding RNA 221
There are no transcripts associated with this gene.

View Full Product Details