Product Details

SNP ID
rs35330772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143599394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCGAGGACAGTGCTGGGCAGCC[G/T]GCCCAGGCCGAGGAAGCCGCCGAGC
Phenotype
MIM: 130592 MIM: 616408
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EEF1D PubMed Links

Gene Details

Gene
EEF1D
Gene Name
eukaryotic translation elongation factor 1 delta
There are no transcripts associated with this gene.

Gene
PYCRL
Gene Name
pyrroline-5-carboxylate reductase-like
There are no transcripts associated with this gene.

Gene
TIGD5
Gene Name
tigger transposable element derived 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032862.4 1491 Silent Mutation CCG,CCT P497P NP_116251.4

View Full Product Details