Product Details

SNP ID
rs17698215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10708499 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAAAACAACGACAACAACCCGTG[C/T]GTCATCAAAGTCATTTCTCAACCAC
Phenotype
MIM: 602378 MIM: 609615
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNM2 PubMed Links

Gene Details

Gene
DNM2
Gene Name
dynamin 2
There are no transcripts associated with this gene.

Gene
MIR638
Gene Name
microRNA 638
There are no transcripts associated with this gene.

Gene
QTRT1
Gene Name
queuine tRNA-ribosyltransferase catalytic subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031209.2 Intron NP_112486.1

View Full Product Details