Product Details

SNP ID
rs17272037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:24057364 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCATTAGTAAGCCTAGCTTATAGG[C/G]AGGTAACAACTCTCATATCTGGACC
Phenotype
MIM: 604768
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF254 PubMed Links

Gene Details

Gene
ZNF254
Gene Name
zinc finger protein 254
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278661.1 Intron NP_001265590.1
NM_001278662.1 Intron NP_001265591.1
NM_001278663.1 Intron NP_001265592.1
NM_001278664.1 Intron NP_001265593.1
NM_001278665.1 Intron NP_001265594.1
NM_001278677.1 Intron NP_001265606.1
NM_001278678.1 Intron NP_001265607.1
NM_203282.3 Intron NP_975011.3
XM_011528443.1 Intron XP_011526745.1
XM_011528444.1 Intron XP_011526746.1
XM_011528448.2 Intron XP_011526750.1
XM_017027514.1 Intron XP_016883003.1
XM_017027515.1 Intron XP_016883004.1
XM_017027516.1 Intron XP_016883005.1
XM_017027517.1 Intron XP_016883006.1
XM_017027518.1 Intron XP_016883007.1
XM_017027519.1 Intron XP_016883008.1

View Full Product Details