Product Details

SNP ID
rs16931334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:64591813 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGTTTCTGCTGAGTGTTTCATTA[C/T]GTTGGTCATTGATATAAAACAATAT
Phenotype
MIM: 613483 MIM: 603711
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BHLHE22 PubMed Links

Gene Details

Gene
BHLHE22
Gene Name
basic helix-loop-helix family member e22
There are no transcripts associated with this gene.

Gene
CYP7B1
Gene Name
cytochrome P450 family 7 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324112.1 6554 Intron NP_001311041.1
NM_004820.4 6554 UTR 3 NP_004811.1
XM_017014002.1 6554 UTR 3 XP_016869491.1

View Full Product Details