Product Details

SNP ID
rs34036869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29112563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGGGAGGGTCACTTAGTGTGT[C/T]AAGGTTGAGACTGTTTTGATTATCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928215 PubMed Links

Gene Details

Gene
LOC101928215
Gene Name
uncharacterized LOC101928215
There are no transcripts associated with this gene.

Gene
RRN3P2
Gene Name
RRN3 homolog, RNA polymerase I transcription factor pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details