Product Details

SNP ID
rs13312800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4368571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCAAATAAATAAATAAATACTG[C/T]CACATGACGAGGGATATAAGAAATT
Phenotype
MIM: 605380 MIM: 610775
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF23 PubMed Links

Gene Details

Gene
FGF23
Gene Name
fibroblast growth factor 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020638.2 2674 UTR 3 NP_065689.1
Gene
TIGAR
Gene Name
TP53 induced glycolysis regulatory phosphatase
There are no transcripts associated with this gene.

View Full Product Details