Product Details

SNP ID
rs17436339
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:202208781 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACTGTCTAGATTGGCATGGGGGC[A/G]CAAAGGCACCATGTTGCGGGGCCCA
Phenotype
MIM: 606653
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LGR6 PubMed Links

Gene Details

Gene
LGR6
Gene Name
leucine rich repeat containing G protein-coupled receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017403.1 Intron NP_001017403.1
NM_001017404.1 Intron NP_001017404.1
NM_021636.2 Intron NP_067649.2
XM_005245404.3 Intron XP_005245461.1
XM_011509838.2 Intron XP_011508140.1
XM_011509839.2 Intron XP_011508141.1
XM_011509840.2 Intron XP_011508142.1
XM_011509841.2 Intron XP_011508143.1
XM_011509842.2 Intron XP_011508144.1
XM_011509843.2 Intron XP_011508145.1
XM_011509844.2 Intron XP_011508146.1
XM_011509846.2 Intron XP_011508148.1
XM_017001996.1 Intron XP_016857485.1
XM_017001997.1 Intron XP_016857486.1
XM_017001998.1 Intron XP_016857487.1

View Full Product Details