Product Details

SNP ID
rs17208939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:24702108 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGCCCCAGGCTGAGCAACATCT[A/G]CAAAAGCTACACAAGACTAGCTGCC
Phenotype
MIM: 608798 MIM: 606959
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DFNA5 PubMed Links

Gene Details

Gene
DFNA5
Gene Name
DFNA5, deafness associated tumor suppressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127453.1 Intron NP_001120925.1
NM_001127454.1 Intron NP_001120926.1
NM_004403.2 Intron NP_004394.1
XM_017011802.1 Intron XP_016867291.1
Gene
MPP6
Gene Name
membrane palmitoylated protein 6
There are no transcripts associated with this gene.

View Full Product Details