Product Details

SNP ID
rs17791935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:34931972 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGACCTGTAGATACATATATCCAG[A/G]TGTTGCGGAAAGAGACACCTCAGTT
Phenotype
MIM: 613892
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPY19L1 PubMed Links

Gene Details

Gene
DPY19L1
Gene Name
dpy-19 like 1 (C. elegans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015283.1 Intron NP_056098.1
XM_011515245.2 Intron XP_011513547.1
XM_011515246.2 Intron XP_011513548.1
Gene
MIR548N
Gene Name
microRNA 548n
There are no transcripts associated with this gene.

View Full Product Details