Product Details

SNP ID
rs17509194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39748196 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCCATTCTGTCCTCCTCCCTTT[A/T]TGGCTAAGCCTCTTCATACTCATGA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00265 PubMed Links

Gene Details

Gene
LINC00265
Gene Name
long intergenic non-protein coding RNA 265
There are no transcripts associated with this gene.

View Full Product Details