Product Details

SNP ID
rs16956303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:55724232 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACCACAGCACACTCTCTCCAGCC[G/T]TGCCTTGTAGCAATACCCTGATGAC
Phenotype
MIM: 616334
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM100 PubMed Links

Gene Details

Gene
TMEM100
Gene Name
transmembrane protein 100
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099640.1 Intron NP_001093110.1
NM_018286.2 Intron NP_060756.2
XM_017024814.1 Intron XP_016880303.1
XM_017024815.1 Intron XP_016880304.1
XM_017024816.1 Intron XP_016880305.1

View Full Product Details