Product Details

SNP ID
rs16948695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50095003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGAAGGGTTAAAAGGAGCCTGC[A/G]GGTGAAGGAGATGCGAGGGGGTAGC
Phenotype
MIM: 605025 MIM: 602525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGA3 PubMed Links

Gene Details

Gene
ITGA3
Gene Name
integrin subunit alpha 3
There are no transcripts associated with this gene.

Gene
PDK2
Gene Name
pyruvate dehydrogenase kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199898.1 Intron NP_001186827.1
NM_001199899.1 Intron NP_001186828.1
NM_001199900.1 Intron NP_001186829.1
NM_002611.4 Intron NP_002602.2

View Full Product Details