Product Details

SNP ID
rs16987899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:27460241 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCTTGCTTTTTTAATGAATGAAA[A/G]TACACTCTGAAAGCTACTTCCTTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP4R3CP PubMed Links

Gene Details

Gene
PPP4R3CP
Gene Name
protein phosphatase 4 regulatory subunit 3C, pseudogene
There are no transcripts associated with this gene.

View Full Product Details