Product Details

SNP ID
rs16971544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:79908265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTTTATTTAGGTAGATACGATAA[A/G]CATTTTAATAGTTACACTTGCGCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ST20 PubMed Links

Gene Details

Gene
ST20
Gene Name
suppressor of tumorigenicity 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100879.1 Intron NP_001094349.1
NM_001100880.2 Intron NP_001094350.1
NM_001199757.1 Intron NP_001186686.1
Gene
ST20-MTHFS
Gene Name
ST20-MTHFS readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199760.1 Intron NP_001186689.1

View Full Product Details