Product Details

SNP ID
rs16969922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78513447 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGGTAAGAGATGACCAATTCGCC[C/G]ATCCATTACCTATCCAGACACATCA
Phenotype
MIM: 614681
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HYKK PubMed Links

Gene Details

Gene
HYKK
Gene Name
hydroxylysine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013619.3 Intron NP_001013641.2
NM_001083612.1 Intron NP_001077081.1
XM_011521231.2 Intron XP_011519533.1
XM_017021922.1 Intron XP_016877411.1

View Full Product Details