Product Details

SNP ID
rs35811768
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73767456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTACATTAATGTAATTTAATAA[A/C]CAAAAGGAAAGAAAATGCAGAATCA
Phenotype
MIM: 603092
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DUSP11 PubMed Links

Gene Details

Gene
DUSP11
Gene Name
dual specificity phosphatase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003584.2 Intron NP_003575.2

View Full Product Details