Product Details

SNP ID
rs17539290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61020375 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCCAGTTGGTTATTAACTGGTA[C/T]GGCAGAATGCTATTGATTTTTTTTA
Phenotype
MIM: 601789 MIM: 612787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PEX13 PubMed Links

Gene Details

Gene
PEX13
Gene Name
peroxisomal biogenesis factor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002618.3 Intron NP_002609.1
Gene
PUS10
Gene Name
pseudouridylate synthase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322123.1 Intron NP_001309052.1
NM_001322124.1 Intron NP_001309053.1
NM_001322127.1 Intron NP_001309056.1
NM_144709.3 Intron NP_653310.2

View Full Product Details