Product Details
- SNP ID
-
rs16941873
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:88460328 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCAGTCGCTATCTGCTGGTGAAGG[A/T]CGAGGACATTTGGTCTGTAAAACAA
- Phenotype
-
MIM: 611851
MIM: 611977
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL46
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16941874] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL46
- Gene Name
- mitochondrial ribosomal protein L46
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022163.3 |
|
Intron |
|
|
NP_071446.2 |
- Gene
- MRPS11
- Gene Name
- mitochondrial ribosomal protein S11
There are no transcripts associated with this gene.
View Full Product Details