Product Details

SNP ID
rs17030027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:153473211 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTTCTTGAGCAGGGACTGAAGTG[A/G]TGACATTGAGTGCTGGGTCCTTACT
Phenotype
MIM: 616243
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0922 PubMed Links

Gene Details

Gene
KIAA0922
Gene Name
KIAA0922
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131007.1 Intron NP_001124479.1
NM_015196.3 Intron NP_056011.3
XM_011531780.1 Intron XP_011530082.1
XM_011531781.1 Intron XP_011530083.1
XM_011531783.1 Intron XP_011530085.1
XM_017007924.1 Intron XP_016863413.1
XM_017007925.1 Intron XP_016863414.1
XM_017007926.1 Intron XP_016863415.1
XM_017007927.1 Intron XP_016863416.1

View Full Product Details