Product Details

SNP ID
rs17106971
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:52029851 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGGACTTCTGCCTGCAATACTTG[C/T]AAAAGCTTGTAAACCAAATGAGCAG
Phenotype
MIM: 609448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KTI12 PubMed Links

Gene Details

Gene
KTI12
Gene Name
KTI12 chromatin associated homolog
There are no transcripts associated with this gene.

Gene
TXNDC12
Gene Name
thioredoxin domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015913.3 Intron NP_056997.1

View Full Product Details