Product Details

SNP ID
rs17742347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43769100 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCGGACCTCAAGACGTGAGAGGG[C/T]TGCCTATGCACAGGGTGTGGAAGGC
Phenotype
MIM: 600183
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf105 PubMed Links

Gene Details

Gene
C17orf105
Gene Name
chromosome 17 open reading frame 105
There are no transcripts associated with this gene.

Gene
DUSP3
Gene Name
dual specificity phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004090.3 1143 UTR 3 NP_004081.1

View Full Product Details