Product Details

SNP ID
rs16851946
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:15691781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTTTTCTACTCCTGAGATAGGT[G/T]AGATCTGCAAGTACAAAAAAATTAG
Phenotype
MIM: 609613
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLEKHM2 PubMed Links

Gene Details

Gene
PLEKHM2
Gene Name
pleckstrin homology and RUN domain containing M2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015164.2 Intron NP_055979.2
XM_005245790.3 Intron XP_005245847.1
XM_005245791.4 Intron XP_005245848.1
XM_017000757.1 Intron XP_016856246.1
XM_017000758.1 Intron XP_016856247.1

View Full Product Details