Product Details
- SNP ID
-
rs17519579
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:31719204 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGGTGCAAGCACGGAGGTATACAGG[A/G]AACTTGGGCTTGATTTGGGACACTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF720
PubMed Links
Gene Details
- Gene
- ZNF720
- Gene Name
- zinc finger protein 720
View Full Product Details