Product Details

SNP ID
rs16831746
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:160289445 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAATGTCTAAGGAAAATGGGAG[A/G]TGATTAAGAGTTGGTGTGGCCTAGT
Phenotype
MIM: 601924 MIM: 600279
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPA PubMed Links

Gene Details

Gene
COPA
Gene Name
coatomer protein complex subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098398.1 4808 UTR 3 NP_001091868.1
NM_004371.3 4808 UTR 3 NP_004362.2
Gene
PEX19
Gene Name
peroxisomal biogenesis factor 19
There are no transcripts associated with this gene.

View Full Product Details