Product Details

SNP ID
rs35774487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55534111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCACACCTTCTAATTAGGAACTA[C/G]TTCTAGCCTTTTATAGCCGAAATAT
Phenotype
MIM: 607124
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PLPP1 PubMed Links

Gene Details

Gene
PLPP1
Gene Name
phospholipid phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003711.3 Intron NP_003702.2
NM_176895.2 Intron NP_795714.1
XM_006714724.2 Intron XP_006714787.1
Gene
RNF138P1
Gene Name
ring finger protein 138 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details