Product Details
- SNP ID
-
rs17626032
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:18389602 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCCTTGACTTTATGACATTACTTT[A/C]TTGGAGAGTGAAGTTTTTAGTGTTT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RNF144B
PubMed Links
Gene Details
- Gene
- RNF144B
- Gene Name
- ring finger protein 144B
View Full Product Details