Product Details

SNP ID
rs17023979
Assay Type
Functionally Tested
NCBI dbSNP Submissions
23
Location
Chr.1:119564860 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTGCACAGGCACACAGCACAGAA[C/T]AGAGATGGCAAGAGGCAGTATGAGG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HSD3BP4 PubMed Links

Gene Details

Gene
HSD3BP4
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta, pseudogene 4
There are no transcripts associated with this gene.

View Full Product Details