Product Details
- SNP ID
-
rs17514413
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:37757976 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAACTAAAGTTTGGCCCAATTTTGA[C/G]CATTTGATGGAGTACAAGAAAAGCA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TRMT10B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117602153] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRMT10B
- Gene Name
- tRNA methyltransferase 10B
View Full Product Details