Product Details
- SNP ID
-
rs7292620
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:31221375 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCGTTAATTTGTATTCATCCTTTAG[A/G]GCTCAGCTTGGAAATTACTTCCTCA
- Phenotype
-
MIM: 601988
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LIMK2
PubMed Links
Gene Details
- Gene
- LIMK2
- Gene Name
- LIM domain kinase 2
View Full Product Details