Product Details

SNP ID
rs17114406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102153618 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAGTTGACTGTTAAGGAGACTGC[A/G]GAGAGTGCTTTCACAGTGGACTGGA
Phenotype
MIM: 602394
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOLC1 PubMed Links

Gene Details

Gene
NOLC1
Gene Name
nucleolar and coiled-body phosphoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284388.1 Intron NP_001271317.1
NM_001284389.1 Intron NP_001271318.1
NM_004741.4 Intron NP_004732.2
XM_005270273.1 Intron XP_005270330.1
Gene
PPRC1
Gene Name
peroxisome proliferator-activated receptor gamma, coactivator-related 1
There are no transcripts associated with this gene.

View Full Product Details