Product Details

SNP ID
rs17722299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100048406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTAGAGCTGTGCAATCACTTAAG[A/G]TTTTGCAAGGGCCAAGGGTGGTGGC
Phenotype
MIM: 603103
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPN1 PubMed Links

Gene Details

Gene
CPN1
Gene Name
carboxypeptidase N subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308.2 Intron NP_001299.1

View Full Product Details