Product Details

SNP ID
rs35108910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150935313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTAACTTGAAGTAGGTTGTAAG[A/C]GGCCAAATATCATATTGTAATCCCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF300P1 PubMed Links

Gene Details

Gene
ZNF300P1
Gene Name
zinc finger protein 300 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details