Product Details

SNP ID
rs17112254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151744695 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGTTTTTATAAAATACAGTCTTAC[A/G]GGAAGCCCAATATGACACAGGACCG
Phenotype
MIM: 602270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATOX1 PubMed Links

Gene Details

Gene
ATOX1
Gene Name
antioxidant 1 copper chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004045.3 Intron NP_004036.1

View Full Product Details