Product Details

SNP ID
rs17854365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23954582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCTTCCAGCTCAGCCTCAAGCC[A/C]TGCCTCTTGTCACAGTGAGGGTTCT
Phenotype
MIM: 602304
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NR1D2 PubMed Links

Gene Details

Gene
NR1D2
Gene Name
nuclear receptor subfamily 1 group D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145425.1 381 UTR 5 NP_001138897.1
NM_005126.4 381 Missense Mutation CAT,CCT H21P NP_005117.3
XM_006713451.3 381 Missense Mutation CAT,CCT H21P XP_006713514.1

View Full Product Details