Product Details

SNP ID
rs17120285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22602718 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCACGCTGTTTCAAGGGATAAGAC[A/G]GTCCCGCTTATTATGTACAGACTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD4 PubMed Links

Gene Details

Gene
ABHD4
Gene Name
abhydrolase domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022060.2 Intron NP_071343.2
XM_005267986.4 Intron XP_005268043.1

View Full Product Details