Product Details

SNP ID
rs17059269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:76358570 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTCACTTGAAGTCCTGGCCTGTG[C/G]TTGTTTCATCTGTTTTTTTGCTCAG
Phenotype
MIM: 615114
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF516 PubMed Links

Gene Details

Gene
ZNF516
Gene Name
zinc finger protein 516
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014643.3 7689 UTR 3 NP_055458.1
XM_011526269.1 7689 UTR 3 XP_011524571.1
XM_011526270.2 7689 UTR 3 XP_011524572.1
XM_011526271.2 7689 UTR 3 XP_011524573.1
XM_011526272.2 7689 UTR 3 XP_011524574.1
XM_011526273.2 7689 UTR 3 XP_011524575.1
XM_011526274.2 7689 UTR 3 XP_011524576.1
XM_011526275.2 7689 UTR 3 XP_011524577.1
XM_011526276.1 7689 UTR 3 XP_011524578.1
XM_011526277.2 7689 Intron XP_011524579.1
XM_017026097.1 7689 UTR 3 XP_016881586.1

View Full Product Details