Product Details
- SNP ID
-
rs17447587
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:105041212 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATAGGAACAACAGGTAAAAAACTG[A/G]CTCCAATTCTTGGTAAACACAAGTT
- Phenotype
-
MIM: 615680
MIM: 147678
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CARD16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75903920] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CARD16
- Gene Name
- caspase recruitment domain family member 16
- Gene
- CASP1
- Gene Name
- caspase 1
There are no transcripts associated with this gene.
View Full Product Details