Product Details

SNP ID
rs74315353
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:7965425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCTTGAAGATGCAAAAAAAGA[C/G]GTTTGTAATCCATACATGGAGTTAT
Phenotype
MIM: 602533
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PARK7 PubMed Links

Gene Details

Gene
PARK7
Gene Name
Parkinsonism associated deglycase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123377.1 Intron NP_001116849.1
NM_007262.4 Intron NP_009193.2
XM_005263424.3 Intron XP_005263481.1

View Full Product Details