Product Details
- SNP ID
-
rs104894267
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:64804789 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGGCCTCGGCCGCCTCGGCCTCTC[G/A]GCTCACTATGCGCACCTTCTGCCGC
- Phenotype
-
MIM: 603166
MIM: 613733
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAP4K2
PubMed Links
Gene Details
- Gene
- MAP4K2
- Gene Name
- mitogen-activated protein kinase kinase kinase kinase 2
There are no transcripts associated with this gene.
- Gene
- MEN1
- Gene Name
- menin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000244.3 |
1870 |
Nonsense Mutation |
CGA,TGA |
R465* |
NP_000235.2 |
NM_130799.2 |
1870 |
Nonsense Mutation |
CGA,TGA |
R460* |
NP_570711.1 |
NM_130800.2 |
1870 |
Nonsense Mutation |
CGA,TGA |
R465* |
NP_570712.1 |
NM_130801.2 |
1870 |
Nonsense Mutation |
CGA,TGA |
R465* |
NP_570713.1 |
NM_130802.2 |
1870 |
Nonsense Mutation |
CGA,TGA |
R465* |
NP_570714.1 |
NM_130803.2 |
1870 |
Nonsense Mutation |
CGA,TGA |
R465* |
NP_570715.1 |
NM_130804.2 |
1870 |
Nonsense Mutation |
CGA,TGA |
R465* |
NP_570716.1 |
XM_005274001.4 |
1870 |
Nonsense Mutation |
CGA,TGA |
R460* |
XP_005274058.1 |
XM_011545040.1 |
1870 |
Nonsense Mutation |
CGA,TGA |
R502* |
XP_011543342.1 |
XM_011545041.2 |
1870 |
Nonsense Mutation |
CGA,TGA |
R502* |
XP_011543343.1 |
XM_011545042.2 |
1870 |
Nonsense Mutation |
CGA,TGA |
R502* |
XP_011543344.1 |
XM_017017765.1 |
1870 |
Nonsense Mutation |
CGA,TGA |
R507* |
XP_016873254.1 |
XM_017017766.1 |
1870 |
Nonsense Mutation |
CGA,TGA |
R507* |
XP_016873255.1 |
XM_017017767.1 |
1870 |
Nonsense Mutation |
CGA,TGA |
R507* |
XP_016873256.1 |
XM_017017768.1 |
1870 |
Nonsense Mutation |
CGA,TGA |
R507* |
XP_016873257.1 |
XM_017017769.1 |
1870 |
Nonsense Mutation |
CGA,TGA |
R460* |
XP_016873258.1 |
XM_017017770.1 |
1870 |
Nonsense Mutation |
CGA,TGA |
R460* |
XP_016873259.1 |
View Full Product Details