Product Details

SNP ID
rs17146441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64895755 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCCACATGGCTGCCGCAGCTGCA[C/T]GCCTGAGACTGACCGCTCCTTGCCT
Phenotype
MIM: 616225 MIM: 610939 MIM: 610941
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATG2A PubMed Links

Gene Details

Gene
ATG2A
Gene Name
autophagy related 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015104.2 Intron NP_055919.2
XM_005273849.3 Intron XP_005273906.1
XM_005273850.3 Intron XP_005273907.1
XM_011544863.2 Intron XP_011543165.1
XM_011544864.2 Intron XP_011543166.1
XM_011544865.2 Intron XP_011543167.1
XM_011544866.2 Intron XP_011543168.1
XM_011544867.2 Intron XP_011543169.1
XM_011544868.2 Intron XP_011543170.1
Gene
MIR192
Gene Name
microRNA 192
There are no transcripts associated with this gene.

Gene
MIR194-2
Gene Name
microRNA 194-2
There are no transcripts associated with this gene.

Gene
MIR194-2HG
Gene Name
MIR194-2 host gene
There are no transcripts associated with this gene.

Gene
MIR6749
Gene Name
microRNA 6749
There are no transcripts associated with this gene.

Gene
MIR6750
Gene Name
microRNA 6750
There are no transcripts associated with this gene.

View Full Product Details