Product Details

SNP ID
rs35780092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74749259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCGACGACGGCCAGGCCGACTCA[A/G]AGGTGCTGGGCGAGTGCGCCCGCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF169 PubMed Links

Gene Details

Gene
RNF169
Gene Name
ring finger protein 169
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098638.1 404 Missense Mutation AAG,GAG K127E NP_001092108.1
XM_011544889.2 404 Missense Mutation AAG,GAG K127E XP_011543191.1

View Full Product Details