Product Details

SNP ID
rs7789085
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.7:100875204 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
CCCACTTTTGTTCGCCTCTCTTCGG[C/T]CCTCTACTCAAGAGCTCCGTCTCCG
Phenotype
MIM: 616861 MIM: 614469 MIM: 602933
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6875 PubMed Links

Gene Details

Gene
MIR6875
Gene Name
microRNA 6875
There are no transcripts associated with this gene.

Gene
SLC12A9
Gene Name
solute carrier family 12 member 9
There are no transcripts associated with this gene.

Gene
SRRT
Gene Name
serrate, RNA effector molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128852.1 126 UTR 5 NP_001122324.1
NM_001128853.1 126 UTR 5 NP_001122325.1
NM_001128854.1 126 UTR 5 NP_001122326.1
NM_015908.5 126 UTR 5 NP_056992.4
XM_005250405.2 126 Intron XP_005250462.1
XM_005250406.2 126 Intron XP_005250463.1
XM_005250407.2 126 Intron XP_005250464.1
XM_005250408.2 126 Intron XP_005250465.1
XM_017012290.1 126 UTR 5 XP_016867779.1
XM_017012291.1 126 UTR 5 XP_016867780.1
Gene
TRIP6
Gene Name
thyroid hormone receptor interactor 6
There are no transcripts associated with this gene.

View Full Product Details