Product Details

SNP ID
rs16964760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52196688 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATGGTGGTCTAGGCTGAGGGGCC[A/G]GAACAGCTACAGACTTTGGGGAAAA
Phenotype
MIM: 604447 MIM: 610022
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNB5 PubMed Links

Gene Details

Gene
GNB5
Gene Name
G protein subunit beta 5
There are no transcripts associated with this gene.

Gene
LOC100129973
Gene Name
uncharacterized LOC100129973
There are no transcripts associated with this gene.

Gene
MYO5C
Gene Name
myosin VC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018728.3 Intron NP_061198.2
XM_011521781.2 Intron XP_011520083.1
XM_017022408.1 Intron XP_016877897.1
XM_017022409.1 Intron XP_016877898.1
XM_017022410.1 Intron XP_016877899.1

View Full Product Details