Product Details

SNP ID
rs16993813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:141618072 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGATCCATAATTGTCTGGTTCCAA[A/G]GATCATTTTCTCAATAATTATCATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC645188 PubMed Links

Gene Details

Gene
LOC645188
Gene Name
uncharacterized LOC645188
There are no transcripts associated with this gene.

Gene
SPANXA2-OT1
Gene Name
SPANXA2 overlapping transcript 1
There are no transcripts associated with this gene.

View Full Product Details