Product Details
- SNP ID
-
rs16854408
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:169819776 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAGCCTGGAGTACATAATCAGTG[C/T]TCTTGGTGTTCCCTATTTTTATTTG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC34
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80069521] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRRC34
- Gene Name
- leucine rich repeat containing 34
There are no transcripts associated with this gene.
- Gene
- LRRIQ4
- Gene Name
- leucine rich repeats and IQ motif containing 4
View Full Product Details