Product Details

SNP ID
rs17089760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:72792522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAACATTGAGATTAGACAACCAA[C/T]TGGCTTTTCAACAGAAAGATGCCAG
Phenotype
MIM: 607532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIBF1 PubMed Links

Gene Details

Gene
PIBF1
Gene Name
progesterone immunomodulatory binding factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006346.2 661 Silent Mutation CTG,TTG L110L NP_006337.2
XM_011534881.2 661 Silent Mutation CTG,TTG L110L XP_011533183.1
XM_011534882.2 661 Silent Mutation CTG,TTG L110L XP_011533184.1
XM_011534884.2 661 Silent Mutation CTG,TTG L110L XP_011533186.1
XM_011534885.2 661 UTR 5 XP_011533187.1
XM_011534886.2 661 Silent Mutation CTG,TTG L110L XP_011533188.1
XM_017020350.1 661 UTR 5 XP_016875839.1
XM_017020351.1 661 Silent Mutation CTG,TTG L110L XP_016875840.1
XM_017020352.1 661 UTR 5 XP_016875841.1
XM_017020353.1 661 UTR 5 XP_016875842.1
XM_017020354.1 661 UTR 5 XP_016875843.1

View Full Product Details