Product Details

SNP ID
rs17100865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:63207414 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACCACCACTCTCCTGGTATATGA[C/T]CTGGTTGAACATAAATCCTTAGGCA
Phenotype
MIM: 607653
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHOJ PubMed Links

Gene Details

Gene
RHOJ
Gene Name
ras homolog family member J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020663.4 Intron NP_065714.1
XM_006720214.3 Intron XP_006720277.1
XM_011536993.2 Intron XP_011535295.1

View Full Product Details