Product Details

SNP ID
rs17807992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92133833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTACCTTATACCACATTTTTTGTA[A/G]CTATGGAGAGCACAGGTAGTAATAA
Phenotype
MIM: 606028
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPSF2 PubMed Links

Gene Details

Gene
CPSF2
Gene Name
cleavage and polyadenylation specific factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322270.1 Intron NP_001309199.1
NM_001322271.1 Intron NP_001309200.1
NM_001322272.1 Intron NP_001309201.1
NM_017437.2 Intron NP_059133.1

View Full Product Details