Product Details

SNP ID
rs17752285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94442658 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCAGGAAGAGTAAGCTCTGGGTG[C/G]TGACCTCCCAAAGGAGCAAGAGGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SERPINA11 PubMed Links

Gene Details

Gene
SERPINA11
Gene Name
serpin family A member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080451.1 832 Missense Mutation ACC,AGC T406S NP_001073920.1
XM_006720105.3 832 Missense Mutation ACC,AGC T202S XP_006720168.1

View Full Product Details